Assay Details
Gene Symbol: AFF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014423.3 | NP_055238.1 | 17 - 18 | 3414 | 62 | |
XM_005271963.4 | XP_005272020.1 | 18 - 19 | 3478 | 62 | ||
XM_005271964.4 | XP_005272021.1 | 11 - 12 | 2126 | 62 | ||
XM_006714587.3 | XP_006714650.1 | 16 - 17 | 3327 | 62 | ||
GenBank mRNA | AF197927.1 | - | 17 - 18 | 3343 | 62 | |
AF213987.2 | - | 17 - 18 | 3071 | 62 | ||
BC137226.1 | - | 17 - 18 | 3312 | 9021223 | 62 | |
BC144538.1 | - | 18 - 19 | 3329 | 62 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05001776.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
