Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023275.hg.1 |
TC04001670.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023275.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023275.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183884.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183884.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183884.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183884.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183884.hg.1 |
TC0400012213.hg.1 |
Clariom D TC |