Assay Details
Gene Symbol: GRTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286732.1 | NP_001273661.1 | 3 - 4 | 442 | 65 | |
NM_001286733.1 | NP_001273662.1 | 3 - 4 | 442 | 65 | ||
NM_024719.3 | NP_078995.2 | 3 - 4 | 442 | 65 | ||
XM_011537521.2 | XP_011535823.1 | 3 - 4 | 438 | 65 | ||
XM_011537522.2 | XP_011535824.1 | 3 - 4 | 438 | 65 | ||
XM_017020755.1 | XP_016876244.1 | 3 - 4 | 400 | 65 | ||
GenBank mRNA | AB449887.1 | - | 3 - 4 | 345 | 65 | |
AK026127.1 | - | 1 - 2 | 85 | 65 | ||
BC033071.2 | - | 1 - 2 | 166 | 65 | ||
BC062461.1 | - | 2 - 3 | 290 | 65 | ||
BC080640.1 | - | 3 - 4 | 399 | 65 | ||
BC112123.1 | - | 1 - 2 | 119 | 8327591 | 65 | |
BC112125.1 | - | 1 - 2 | 131 | 8327593 | 65 | |
BC143682.1 | - | 1 - 2 | 119 | 65 | ||
BQ689859.1 | - | 3 - 4 | 379 | 65 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000893.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR13010008.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13010008.hg.1 | ENST00000375430.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13010008.hg.1 | ENST00000375431.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13010008.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13010008.hg.1 | TC13000893.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000476439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000476439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000476439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165919.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165919.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165919.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165919.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165919.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165919.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165919.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165919.hg.1 | TC1300009898.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300165919.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165919.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165919.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165919.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165919.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165919.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165919.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000476439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000476439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000476439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
