Assay Details
Gene Symbol: DESI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297746.1 | NP_001284675.1 | 1 - 2 | 302 | 124 | |
NM_016076.4 | NP_057160.2 | 1 - 2 | 302 | 124 | ||
NR_123735.1 | 1 - 2 | 302 | 124 | |||
NR_123736.1 | 1 - 2 | 302 | 124 | |||
GenBank mRNA | AF151904.1 | - | 1 - 2 | 94 | 124 | |
AK314664.1 | - | 1 - 2 | 272 | 124 | ||
BC004485.2 | - | 1 - 2 | 249 | 3831754 | 124 | |
BI459676.1 | - | 1 - 2 | 306 | 123 | ||
CN303012.1 | - | 1 - 2 | 213 | 124 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001989.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900011558.hg.1 | ENST00000596280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030820.hg.1 | ENST00000302550.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030820.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01030820.hg.1 | TC01001989.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01030828.hg.1 | ENST00000302550.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030828.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01030828.hg.1 | TC01001989.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01030823.hg.1 | ENST00000263831.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030823.hg.1 | ENST00000302550.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030823.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030823.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01030823.hg.1 | TC01001989.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01030827.hg.1 | ENST00000263831.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030827.hg.1 | ENST00000302550.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030827.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01030827.hg.1 | TC01001989.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190005.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190005.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190005.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190005.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190005.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190005.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190005.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190005.hg.1 | TC0100012222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100190005.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190005.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190005.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190005.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190005.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190005.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190005.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189997.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189997.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189997.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189997.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189997.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189997.hg.1 | TC0100012222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100189997.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189997.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189997.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189997.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189997.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011558.hg.1 | ENST00000596280.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190006.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190006.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190006.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190006.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190006.hg.1 | TC0100012222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100190006.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190006.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190006.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190006.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190000.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190000.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190000.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190000.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190000.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190000.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190000.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190000.hg.1 | TC0100012222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100190000.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190000.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190000.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190000.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190000.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190000.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190000.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190001.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190001.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190001.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190001.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190001.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190001.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190001.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190001.hg.1 | TC0100012222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100190001.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190001.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190001.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190001.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190001.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190001.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190001.hg.1 | NR_123736.1 | Refseq |
