Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011222.hg.1 |
ENST00000326134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011222.hg.1 |
ENST00000398656.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011222.hg.1 |
ENST00000420663.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011222.hg.1 |
ENST00000528919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011222.hg.1 |
ENST00000534366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011222.hg.1 |
ENST00000544576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011222.hg.1 |
NM_032450.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011222.hg.1 |
TC08000844.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0800163239.hg.1 |
ENST00000326134.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163239.hg.1 |
ENST00000527552.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163239.hg.1 |
ENST00000528919.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163239.hg.1 |
ENST00000532255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163239.hg.1 |
ENST00000534366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163239.hg.1 |
ENST00000544576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163239.hg.1 |
NM_001288814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800163239.hg.1 |
NM_032450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800163239.hg.1 |
TC0800009244.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800163239.hg.1 |
ENST00000326134.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163239.hg.1 |
ENST00000527552.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163239.hg.1 |
ENST00000528919.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163239.hg.1 |
ENST00000532255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163239.hg.1 |
ENST00000534366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163239.hg.1 |
ENST00000544576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163239.hg.1 |
NM_001288814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800163239.hg.1 |
NM_032450.2 |
Refseq |