Assay Details
Gene Symbol: GRTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024719.3 | NP_078995.2 | 7 - 8 | 1016 | 75 | |
GenBank mRNA | BC033071.2 | - | 5 - 6 | 740 | 75 | |
BC062461.1 | - | 6 - 7 | 864 | 75 | ||
BC080640.1 | - | 7 - 8 | 973 | 75 | ||
CB242314.1 | - | 1 - 2 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000893.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR13010000.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13010000.hg.1 | TC13000893.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13010001.hg.1 | ENST00000375431.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13010001.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13010001.hg.1 | TC13000893.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000476439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000476439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000476439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165909.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165909.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165909.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165909.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165909.hg.1 | TC1300009898.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300165909.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165909.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165909.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165909.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165910.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165910.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165910.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165910.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165910.hg.1 | TC1300009898.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300165910.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165910.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165910.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165910.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165911.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165911.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165911.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165911.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165911.hg.1 | TC1300009898.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300165911.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165911.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165911.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165911.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000326039.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000375431.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000476439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000476439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000476439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009898.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165908.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165908.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165908.hg.1 | NM_024719.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165908.hg.1 | TC1300009898.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300165908.hg.1 | ENST00000620217.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165908.hg.1 | NM_001286733.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165908.hg.1 | NM_024719.3 | Refseq |
