Assay Details
Gene Symbol: GRIK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175768.3 | NP_786944.1 | 16 - 17 | 2942 | 101 | |
GenBank mRNA | AJ301610.1 | - | 16 - 17 | 2649 | 101 | |
HM149336.1 | - | 16 - 17 | 2649 | 101 | ||
HM149338.1 | - | 13 - 14 | 2088 | 101 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000836.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
