Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000740.hg.1 |
TC01000053.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000740.hg.1 |
ENST00000378585.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000740.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006548.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100145241.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100145241.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100145241.hg.1 |
ENST00000378585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100145241.hg.1 |
NM_000815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100145241.hg.1 |
TC0100006548.hg.1 |
Clariom D TC |