Assay Details
Gene Symbol: MROH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001288814.1 | NP_001275743.1 | 40 - 41 | 4870 | 140 | |
NM_032450.2 | NP_115826.2 | 42 - 43 | 5010 | 140 | ||
XM_011517270.2 | XP_011515572.1 | 43 - 44 | 5202 | 140 | ||
XM_011517272.2 | XP_011515574.1 | 42 - 43 | 5175 | 140 | ||
XM_011517273.2 | XP_011515575.1 | 42 - 43 | 5146 | 140 | ||
XM_011517274.2 | XP_011515576.1 | 41 - 42 | 5119 | 140 | ||
XM_011517275.2 | XP_011515577.1 | 41 - 42 | 4957 | 140 | ||
XM_011517276.2 | XP_011515578.1 | 43 - 44 | 5228 | 140 | ||
XM_011517277.1 | XP_011515579.1 | 43 - 44 | 5064 | 140 | ||
XM_011517278.1 | XP_011515580.1 | 42 - 43 | 4944 | 140 | ||
XM_011517279.2 | XP_011515581.1 | 42 - 43 | 5108 | 140 | ||
XM_011517281.1 | XP_011515583.1 | 42 - 43 | 5117 | 140 | ||
XM_011517282.1 | XP_011515584.1 | 41 - 42 | 5042 | 140 | ||
XM_011517283.2 | XP_011515585.1 | 43 - 44 | 5023 | 140 | ||
XM_017013803.1 | XP_016869292.1 | 40 - 41 | 4929 | 140 | ||
XM_017013804.1 | XP_016869293.1 | 42 - 43 | 5037 | 140 | ||
XM_017013805.1 | XP_016869294.1 | 41 - 42 | 4917 | 140 | ||
XM_017013806.1 | XP_016869295.1 | 40 - 41 | 4679 | 140 | ||
GenBank mRNA | AI660468.1 | - | 2 - 3 | 140 | ||
AL834309.1 | - | 41 - 42 | 4894 | 140 | ||
BC017118.1 | - | 10 - 11 | 1427 | 160 | ||
CN333563.1 | - | 3 - 4 | 364 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
