Assay Details
Gene Symbol: MROH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011517282.1 | XP_011515584.1 | 2 - 3 | 384 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011517282.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
