Assay Details
Gene Symbol: EPHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005232.4 | NP_005223.4 | 4 - 5 | 920 | 76 | |
XM_006715880.3 | XP_006715943.1 | 4 - 5 | 893 | 76 | ||
GenBank mRNA | AK290351.1 | - | 4 - 5 | 884 | 76 | |
AK313263.1 | - | 4 - 5 | 893 | 76 | ||
BC130291.1 | - | 4 - 5 | 904 | 40146322 | 76 | |
BC144156.1 | - | 4 - 5 | 904 | 76 | ||
EU826604.1 | - | 4 - 5 | 833 | 76 | ||
EU826605.1 | - | 4 - 5 | 833 | 76 | ||
M18391.1 | - | 4 - 5 | 926 | 76 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001971.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
positive regulation of cell-matrix adhesion
negative regulation of protein kinase activity
cell surface receptor signaling pathway
positive regulation of cell population proliferation
peptidyl-tyrosine phosphorylation
positive regulation of cell migration
negative regulation of cell migration
substrate adhesion-dependent cell spreading
somatic stem cell population maintenance
regulation of GTPase activity
positive regulation of angiogenesis
protein autophosphorylation
ephrin receptor signaling pathway
positive regulation of stress fiber assembly
activation of GTPase activity
protein kinase activity
transmembrane-ephrin receptor activity
ATP binding
protein kinase binding
angiogenesis
positive regulation of cell-matrix adhesion
negative regulation of protein kinase activity
cell surface receptor signaling pathway
positive regulation of cell population proliferation
peptidyl-tyrosine phosphorylation
positive regulation of cell migration
negative regulation of cell migration
substrate adhesion-dependent cell spreading
somatic stem cell population maintenance
regulation of GTPase activity
positive regulation of angiogenesis
protein autophosphorylation
ephrin receptor signaling pathway
positive regulation of stress fiber assembly
activation of GTPase activity
protein kinase activity
transmembrane-ephrin receptor activity
ATP binding
protein kinase binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07030395.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07030395.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07030395.hg.1 | TC07001971.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000458129.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000458129.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000465208.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000465208.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000465208.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000465208.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000465208.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000479459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000479459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000479459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000479459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000479459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000494989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000494989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000494989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000494989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000494989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000497891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000497891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | ENST00000497891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700193004.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700193004.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700193004.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700193004.hg.1 | TC0700012886.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700193004.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700193004.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700193004.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000275815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000458129.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000458129.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000465208.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000465208.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000465208.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000465208.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000465208.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000479459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000479459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000479459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000479459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000479459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000488068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000494989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000494989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000494989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000494989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000494989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000497891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000497891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | ENST00000497891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012886.hg.1 | NM_005232.4 | Refseq |
