Assay Details
Gene Symbol: GNA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002067.4 | NP_002058.2 | 2 - 3 | 570 | 103 | |
GenBank mRNA | AB209435.1 | - | 2 - 3 | 322 | 103 | |
AF011497.1 | - | 2 - 3 | 328 | 103 | ||
AF493900.1 | - | 2 - 3 | 328 | 103 | ||
BC089041.1 | - | 2 - 3 | 570 | 6085072 | 103 | |
BC096225.1 | - | 2 - 3 | 378 | 40005016 | 103 | |
BC096226.3 | - | 2 - 3 | 378 | 40005021 | 103 | |
BC096227.1 | - | 2 - 3 | 378 | 40005025 | 103 | |
CR457004.1 | - | 2 - 3 | 328 | 103 | ||
M69013.1 | - | 2 - 3 | 514 | 103 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000063.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
action potential
signal transduction
G protein-coupled acetylcholine receptor signaling pathway
heart development
phototransduction, visible light
entrainment of circadian clock
platelet activation
regulation of melanocyte differentiation
developmental pigmentation
phospholipase C-activating dopamine receptor signaling pathway
cellular response to pH
GTPase activity
GTP binding
G-protein beta/gamma-subunit complex binding
type 2A serotonin receptor binding
metal ion binding
skeletal system development
action potential
signal transduction
G protein-coupled acetylcholine receptor signaling pathway
heart development
phototransduction, visible light
entrainment of circadian clock
platelet activation
regulation of melanocyte differentiation
developmental pigmentation
phospholipase C-activating dopamine receptor signaling pathway
cellular response to pH
GTPase activity
GTP binding
G-protein beta/gamma-subunit complex binding
type 2A serotonin receptor binding
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19000997.hg.1 | ENST00000078429.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19000997.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19000997.hg.1 | TC19000063.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19000998.hg.1 | ENST00000078429.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19000998.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19000998.hg.1 | TC19000063.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0700011361.hg.1 | ENST00000474858.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011361.hg.1 | ENST00000474858.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011361.hg.1 | ENST00000474858.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000586763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000586763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000586763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000588401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000588401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000588401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000591301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000591301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000591301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900146208.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146208.hg.1 | ENST00000586763.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146208.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146208.hg.1 | ENST00000588401.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146208.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900146208.hg.1 | TC1900006616.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900146208.hg.1 | ENST00000588401.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146208.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146208.hg.1 | ENST00000586763.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146208.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146208.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000586763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000586763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000586763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000588401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000588401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000588401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000591301.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000591301.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000591301.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900146205.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146205.hg.1 | ENST00000586763.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146205.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900146205.hg.1 | TC1900006616.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900146205.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146205.hg.1 | ENST00000586763.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146205.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011361.hg.1 | ENST00000474858.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011361.hg.1 | ENST00000474858.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011361.hg.1 | ENST00000474858.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146209.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146209.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146209.hg.1 | ENST00000588401.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146209.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900146209.hg.1 | TC1900006616.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900146209.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146209.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146209.hg.1 | ENST00000588401.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900146209.hg.1 | NM_002067.4 | Refseq |
