Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034320.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034320.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034320.hg.1 |
TC01002246.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034319.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034319.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034319.hg.1 |
TC01002246.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013003.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100195850.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100195850.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100195850.hg.1 |
TC0100013003.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100195850.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100195850.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100195849.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100195849.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100195849.hg.1 |
TC0100013003.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100195849.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100195849.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100195848.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100195848.hg.1 |
NM_024758.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100195848.hg.1 |
TC0100013003.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100195848.hg.1 |
ENST00000375826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100195848.hg.1 |
NM_024758.4 |
Refseq |