Assay Details
Gene Symbol: COL6A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278298.1 | NP_001265227.1 | 10 - 11 | 4488 | 100 | |
NM_153264.6 | NP_694996.5 | 10 - 11 | 4488 | 100 | ||
NR_022012.2 | 10 - 11 | 4488 | 100 | |||
XM_011512621.2 | XP_011510923.1 | 10 - 11 | 4362 | 100 | ||
XM_011512622.2 | XP_011510924.1 | 10 - 11 | 4362 | 100 | ||
XM_011512623.2 | XP_011510925.1 | 10 - 11 | 4362 | 100 | ||
GenBank mRNA | AM906080.1 | - | 4 - 5 | 1315 | 100 | |
AM906081.1 | - | 2 - 3 | 430 | 100 | ||
EU085556.1 | - | 10 - 11 | 4488 | 100 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000704.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
