Assay Details
Gene Symbol: BRD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007371.3 | NP_031397.1 | 3 - 4 | 546 | 82 | |
XM_006717291.2 | XP_006717354.1 | 3 - 4 | 678 | 82 | ||
XM_011519052.2 | XP_011517354.1 | 3 - 4 | 897 | 82 | ||
XM_017015165.1 | XP_016870654.1 | 2 - 3 | 471 | 82 | ||
GenBank mRNA | AK299519.1 | - | 3 - 4 | 678 | 82 | |
AY513270.1 | - | 2 - 3 | 376 | 82 | ||
BC032124.2 | - | 3 - 4 | 656 | 4856840 | 82 | |
BI545861.1 | - | 3 - 4 | 556 | 82 | ||
D26362.1 | - | 3 - 4 | 497 | 82 | ||
KR633047.1 | - | 2 - 3 | 358 | 82 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001697.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09021842.hg.1 | ENST00000303407.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021842.hg.1 | ENST00000357885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021842.hg.1 | ENST00000371834.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021842.hg.1 | ENST00000540795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021842.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09021842.hg.1 | TC09001697.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181349.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181349.hg.1 | ENST00000371834.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181349.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181349.hg.1 | NM_007371.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181349.hg.1 | TC0900011872.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900181349.hg.1 | ENST00000303407.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181349.hg.1 | ENST00000371834.6 | Ensembl |
