Assay Details
Gene Symbol: RAD51C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058216.2 | NP_478123.1 | 3 - 4 | 644 | 123 | |
XM_006722001.3 | XP_006722064.1 | 3 - 4 | 615 | 123 | ||
XM_006722002.3 | XP_006722065.1 | 3 - 4 | 615 | 123 | ||
XM_006722004.3 | XP_006722067.1 | 3 - 4 | 537 | 123 | ||
XM_006722005.3 | XP_006722068.1 | 3 - 4 | 523 | 123 | ||
XM_011525092.2 | XP_011523394.1 | 3 - 4 | 743 | 123 | ||
XM_011525093.2 | XP_011523395.1 | 3 - 4 | 904 | 123 | ||
XM_011525094.2 | XP_011523396.1 | 3 - 4 | 592 | 123 | ||
XM_017024914.1 | XP_016880403.1 | 3 - 4 | 537 | 123 | ||
XM_017024915.1 | XP_016880404.1 | 3 - 4 | 889 | 123 | ||
XM_017024916.1 | XP_016880405.1 | 4 - 5 | 797 | 123 | ||
XM_017024917.1 | XP_016880406.1 | 3 - 4 | 522 | 123 | ||
XM_017024918.1 | XP_016880407.1 | 3 - 4 | 591 | 123 | ||
XM_017024919.1 | XP_016880408.1 | 3 - 4 | 904 | 123 | ||
GenBank mRNA | AF029669.1 | - | 3 - 4 | 615 | 123 | |
AK303363.1 | - | 3 - 4 | 554 | 123 | ||
BC107753.1 | - | 3 - 4 | 648 | 6715697 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
