Assay Details
Gene Symbol: NCKAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205842.2 | NP_995314.1 | 2 - 3 | 880 | 136 | |
XM_006712200.3 | XP_006712263.1 | 2 - 3 | 372 | 136 | ||
GenBank mRNA | AB011159.1 | - | 2 - 3 | 880 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
