Assay Details
Gene Symbol: PDZRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015009.2 | NP_055824.1 | 3 - 4 | 1011 | 70 | |
XM_017005942.1 | XP_016861431.1 | 2 - 3 | 1006 | 70 | ||
GenBank mRNA | AB029018.1 | - | 3 - 4 | 1011 | 70 | |
BC150623.1 | - | 3 - 4 | 998 | 9056993 | 70 | |
BC152417.1 | - | 3 - 4 | 1034 | 8862525 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
