Assay Details
Gene Symbol: AKAP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178813.5 | NP_848928.1 | 3 - 4 | 397 | 85 | |
GenBank mRNA | AF514780.1 | - | 3 - 4 | 344 | 85 | |
BC066357.1 | - | 4 - 5 | 534 | 5295545 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000590.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X007784.hg.1 | TC0X000590.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007784.hg.1 | ENST00000371431.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007784.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371422.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371422.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371422.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371422.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000491105.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000491105.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000491105.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000491105.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371422.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371422.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371422.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371422.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000491105.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000491105.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000491105.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000491105.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00156473.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00156473.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00156473.hg.1 | ENST00000371431.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00156473.hg.1 | NM_178813.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00156473.hg.1 | TC0X00008264.hg.1 | Clariom D TC |
