Assay Details
Gene Symbol: CDCA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018101.3 | NP_060571.1 | 1 - 2 | 183 | 75 | |
GenBank mRNA | AK001330.1 | - | 1 - 2 | 103 | 75 | |
AK222549.1 | - | 1 - 2 | 103 | 75 | ||
AK222568.1 | - | 1 - 2 | 57 | 75 | ||
AY508815.2 | - | 1 - 2 | 103 | 75 | ||
BC016944.1 | - | 1 - 2 | 56 | 3457122 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000476.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK001330.1, AY508815.2, BC016944.1, NM_018101.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01007402.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01007402.hg.1 | ENST00000373055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01007402.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01007402.hg.1 | TC01000476.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01007400.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01007400.hg.1 | ENST00000373055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01007400.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01007400.hg.1 | TC01000476.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01007404.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01007404.hg.1 | ENST00000373055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01007404.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01007404.hg.1 | TC01000476.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0700011158.hg.1 | ENST00000443044.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155217.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155217.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155217.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155217.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155217.hg.1 | TC0100007841.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100155217.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155217.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155217.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155217.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007841.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155209.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155209.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155209.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155209.hg.1 | TC0100007841.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100155209.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155209.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155209.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155214.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155214.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155214.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155214.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155214.hg.1 | TC0100007841.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100155214.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155214.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155214.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155214.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011158.hg.1 | ENST00000443044.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155218.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155218.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155218.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155218.hg.1 | NM_018101.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155218.hg.1 | TC0100007841.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100155218.hg.1 | ENST00000327331.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155218.hg.1 | ENST00000373055.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155218.hg.1 | NM_001256875.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155218.hg.1 | NM_018101.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 130297 | - | - |
