Assay Details
Gene Symbol: SERPINB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002639.4 | NP_002630.2 | 4 - 5 | 568 | 82 | |
GenBank mRNA | AK301667.1 | - | 3 - 4 | 332 | 82 | |
AK312765.1 | - | 4 - 5 | 546 | 82 | ||
AL602439.1 | - | 4 - 5 | 571 | 82 | ||
BC020713.1 | - | 4 - 5 | 521 | 4248249 | 82 | |
CV575332.1 | - | 1 - 2 | 55 | 82 | ||
U04313.1 | - | 4 - 5 | 501 | 82 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000226.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
morphogenesis of an epithelium
movement of cell or subcellular component
negative regulation of endopeptidase activity
extracellular matrix organization
regulation of epithelial cell proliferation
prostate gland morphogenesis
serine-type endopeptidase inhibitor activity
protein binding
morphogenesis of an epithelium
movement of cell or subcellular component
negative regulation of endopeptidase activity
extracellular matrix organization
regulation of epithelial cell proliferation
prostate gland morphogenesis
serine-type endopeptidase inhibitor activity
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR18002593.hg.1 | ENST00000382771.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18002593.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18002593.hg.1 | TC18000226.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000424602.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000424602.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000424602.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000424602.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000464346.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000464346.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000464346.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000464346.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000465652.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000465652.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000465652.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000465652.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000588986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000588986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000588986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | ENST00000588986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18002597.hg.1 | ENST00000382771.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18002597.hg.1 | ENST00000464346.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18002597.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18002597.hg.1 | TC18000226.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1800151753.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151753.hg.1 | ENST00000464346.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151753.hg.1 | ENST00000465652.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151753.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151753.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800151753.hg.1 | TC1800007543.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800151753.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151753.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151753.hg.1 | ENST00000464346.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151753.hg.1 | ENST00000465652.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151753.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000424602.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000424602.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000424602.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000424602.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000464346.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000464346.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000464346.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000464346.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000465652.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000465652.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000465652.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000465652.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000588986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000588986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000588986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | ENST00000588986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007543.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800151748.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151748.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151748.hg.1 | ENST00000588986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151748.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800151748.hg.1 | TC1800007543.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800151748.hg.1 | ENST00000382771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151748.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151748.hg.1 | ENST00000588986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151748.hg.1 | NM_002639.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800151754.hg.1 | ENST00000489441.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151754.hg.1 | TC1800007543.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800151754.hg.1 | ENST00000489441.5 | Ensembl |
