Assay Details
Gene Symbol: PSMB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002801.3 | NP_002792.1 | 1 - 2 | 187 | 93 | |
GenBank mRNA | AK225658.1 | - | 1 - 2 | 160 | 93 | |
AK312208.1 | - | 1 - 2 | 147 | 93 | ||
BC017198.1 | - | 1 - 2 | 138 | 3143189 | 93 | |
BC052369.1 | - | 1 - 2 | 125 | 4054165 | 93 | |
BM845639.1 | - | 1 - 2 | 160 | 93 | ||
BP289065.1 | - | 1 - 2 | 187 | 93 | ||
Y13640.1 | - | 1 - 2 | 147 | 93 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16002086.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK312208.1,BC017198.1,BC052369.1,NM_002801.3,Y13640.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
protein polyubiquitination
cell morphogenesis
stimulatory C-type lectin receptor signaling pathway
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
regulation of cellular amino acid metabolic process
humoral immune response
viral process
anaphase-promoting complex-dependent catabolic process
tumor necrosis factor-mediated signaling pathway
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
T cell proliferation
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of mRNA stability
T cell receptor signaling pathway
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
Wnt signaling pathway, planar cell polarity pathway
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
threonine-type endopeptidase activity
protein binding
MAPK cascade
protein polyubiquitination
cell morphogenesis
stimulatory C-type lectin receptor signaling pathway
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
regulation of cellular amino acid metabolic process
humoral immune response
viral process
anaphase-promoting complex-dependent catabolic process
tumor necrosis factor-mediated signaling pathway
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
T cell proliferation
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of mRNA stability
T cell receptor signaling pathway
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
Wnt signaling pathway, planar cell polarity pathway
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
threonine-type endopeptidase activity
protein binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR16016704.hg.1 | TC16002086.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16016704.hg.1 | ENST00000358514.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16016704.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16016706.hg.1 | TC16002086.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16016706.hg.1 | ENST00000358514.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16016706.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16016703.hg.1 | TC16002086.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16016703.hg.1 | ENST00000358514.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16016703.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000570985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000570985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000570985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000570985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000570985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000570985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000574576.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000574576.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000574576.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000574576.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000574576.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194025.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194025.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194025.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194025.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194025.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194025.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194025.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194025.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194025.hg.1 | TC1600011553.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600194026.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194026.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194026.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194026.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194026.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194026.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194026.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194026.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194026.hg.1 | TC1600011553.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000570985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000570985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000570985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000570985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000570985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000570985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000574576.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000574576.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000574576.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000574576.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000574576.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011553.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194023.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194023.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194023.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194023.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194023.hg.1 | ENST00000358514.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194023.hg.1 | ENST00000570304.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194023.hg.1 | ENST00000575556.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194023.hg.1 | NM_002801.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194023.hg.1 | TC1600011553.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 156119 | - | - |
