Assay Details
Gene Symbol: DIO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242502.2 | NP_001229431.1 | 2 - 3 | 487 | 144 | |
GenBank mRNA | AB041844.1 | - | 2 - 3 | 173 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000422005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000422005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000422005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000422005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000422005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000422005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000422005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000422005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000422005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000422005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000422005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000422005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000422005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000422005.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000438257.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000438257.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000438257.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000438257.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000438257.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000438257.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000438257.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000438257.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000438257.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000438257.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000438257.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000438257.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000438257.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000553968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000553968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000555750.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000555750.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000555750.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000555750.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000555750.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000555750.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000555750.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000555750.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000555750.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000555750.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000555844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000555844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000555844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000555844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000555844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000556811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000556811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000556811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000556811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000556811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000556811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000556811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000556811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557010.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557010.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557010.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557010.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557010.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557010.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557010.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557010.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557010.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557010.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557010.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557010.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557010.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557010.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557125.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557125.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557125.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557125.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | ENST00000557125.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_000793.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_000793.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_000793.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_000793.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_000793.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_000793.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_000793.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_000793.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_000793.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_000793.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_000793.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_000793.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_000793.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_000793.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001007023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001007023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001007023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001007023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001007023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001007023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001007023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001007023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001007023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001007023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001007023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001007023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001007023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242502.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242502.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242502.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242502.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242502.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242502.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242502.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242502.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242502.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242502.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242502.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242502.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242502.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242502.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242503.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242503.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242503.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242503.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242503.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242503.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242503.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242503.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242503.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242503.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242503.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242503.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_001242503.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_013989.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_013989.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_013989.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_013989.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_013989.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_013989.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_013989.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_013989.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_013989.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_013989.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_013989.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_013989.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010772.hg.1 | NM_013989.4 | Refseq |
