Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007430.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004746.hg.1 |
ENST00000398859.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004746.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004746.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004746.hg.1 |
TC16000331.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004744.hg.1 |
ENST00000398859.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004744.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004744.hg.1 |
TC16000331.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600153857.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153857.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153857.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153857.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153857.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153857.hg.1 |
TC1600007430.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600153857.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153857.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153857.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153857.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153857.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
ENST00000565014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007430.hg.1 |
NR_024370.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153854.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153854.hg.1 |
NR_015396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153854.hg.1 |
TC1600007430.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600153854.hg.1 |
ENST00000398859.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153854.hg.1 |
NR_015396.1 |
Refseq |