Assay Details
Gene Symbol: SDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144952.1 | NP_001138424.1 | 6 - 7 | 723 | 75 | |
XM_011524914.2 | XP_011523216.1 | 6 - 7 | 1107 | 75 | ||
XM_011524915.2 | XP_011523217.1 | 6 - 7 | 1109 | 75 | ||
XM_011524916.2 | XP_011523218.1 | 6 - 7 | 1117 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001847.hg.1
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000585283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000585283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
