Assay Details
Gene Symbol: TRAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032271.2 | NP_115647.2 | 11 - 12 | 1185 | 51 | |
XM_005255627.4 | XP_005255684.1 | 11 - 12 | 1225 | 51 | ||
XM_011522700.1 | XP_011521002.1 | 11 - 12 | 1227 | 51 | ||
GenBank mRNA | AK090624.1 | - | 11 - 12 | 1199 | 51 | |
AL136921.1 | - | 10 - 11 | 1081 | 51 | ||
AY569455.1 | - | 10 - 11 | 1138 | 51 | ||
BC024267.1 | - | 9 - 10 | 971 | 51 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000063.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPKKK activity
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
protein ubiquitination
positive regulation of MAPK cascade
positive regulation of apoptotic signaling pathway
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
activation of MAPKKK activity
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
protein ubiquitination
positive regulation of MAPK cascade
positive regulation of apoptotic signaling pathway
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR16001306.hg.1 | ENST00000326181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16001306.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16001306.hg.1 | TC16000063.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600146342.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600146342.hg.1 | ENST00000570169.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600146342.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600146342.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600146342.hg.1 | ENST00000570169.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600146342.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600146342.hg.1 | TC1600006594.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
