Assay Details
Gene Symbol: DCAF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024345.4 | NP_077321.3 | 1 - 2 | 615 | 72 | |
XM_005251577.3 | XP_005251634.1 | 1 - 2 | 636 | 72 | ||
XM_005251578.3 | XP_005251635.1 | 1 - 2 | 636 | 72 | ||
XM_017015128.1 | XP_016870617.1 | 1 - 2 | 636 | 72 | ||
GenBank mRNA | AK074532.1 | - | 1 - 2 | 102 | 72 | |
AK315756.1 | - | 1 - 2 | 68 | 72 | ||
BC108686.1 | - | 1 - 2 | 584 | 6157119 | 72 | |
BC139834.1 | - | 1 - 2 | 102 | 72 | ||
CD299973.1 | - | 1 - 2 | 130 | 72 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000218.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000452964.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09002589.hg.1 | ENST00000242323.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002589.hg.1 | ENST00000377724.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002589.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09002589.hg.1 | TC09000218.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09002591.hg.1 | ENST00000242323.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002591.hg.1 | ENST00000377724.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002591.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09002591.hg.1 | TC09000218.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0900148908.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148908.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148908.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148908.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148908.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148908.hg.1 | TC0900007167.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900148908.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148908.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148908.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148908.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148908.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000452964.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148904.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148904.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148904.hg.1 | TC0900007167.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900148904.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148904.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148903.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148903.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148903.hg.1 | TC0900007167.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900148903.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148903.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148905.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148905.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148905.hg.1 | TC0900007167.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900148905.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148905.hg.1 | NM_024345.4 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 176130 | - | - |
