Assay Details
Gene Symbol: CPS1-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002763.2 | 1 | 1730 | 137 | ||
GenBank mRNA | AF090897.1 | - | 1 | 1739 | 137 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | LanC like 1 | LANCL1 |
GPR69A, p40 |
-141.2kb | Anti-sense | Chr. 2 - 210431249 - 210477618 on NCBI Build GRCh38 |
Downstream | erb-b2 receptor tyrosine kinase 4 | ERBB4 |
ALS19, HER4, p180erbB4 |
756.1kb | Anti-sense | Chr. 2 - 211375717 - 212539147 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P16694 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P16694 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 358 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 9681 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P16694 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P16694 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857659 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982896 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982920 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982894 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982919 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982935 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982921 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857645 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857674 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857615 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15378683 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857660 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857642 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982918 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15378676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857630 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982899 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857604 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857647 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857596 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982897 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857644 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982922 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15378686 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982886 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857661 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982931 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982888 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982892 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857610 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982930 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982872 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982873 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982943 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857648 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982947 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857601 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857675 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982871 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15378685 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857638 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982882 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982878 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982887 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857650 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982875 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982946 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982876 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982884 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982904 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15378688 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982926 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982945 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15378682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982890 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857671 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982889 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857612 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857665 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982940 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982900 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15378674 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982923 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982891 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857596 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982899 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857604 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982940 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982884 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982943 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982930 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857675 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982900 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982871 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15378685 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857665 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857638 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982920 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982918 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982889 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857612 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857615 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982904 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982887 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982923 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982876 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857674 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982890 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857661 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15378683 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857647 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857642 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982888 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857650 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857659 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982892 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982931 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982945 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982946 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857660 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982886 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15378676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982872 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857601 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982894 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15378688 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982875 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857645 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15378674 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982882 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982891 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857648 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982921 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982922 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982935 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982897 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857671 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857644 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982919 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982926 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982947 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857630 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15378682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01857610 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982873 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982896 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21982878 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15378686 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P16694 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P16694 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857592 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857606 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15378673 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857637 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857656 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857676 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15378688 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857681 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857623 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857632 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857662 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857644 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15378692 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857614 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857634 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857616 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857667 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857596 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15378684 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857649 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857621 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857599 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857652 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857673 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857625 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857664 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857601 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857611 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857658 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857609 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857630 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15378676 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15378690 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857654 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857646 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857635 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15378685 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15378689 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857666 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857617 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857639 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857589 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857659 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15378674 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857618 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857604 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15378679 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857627 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01857670 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NR_002763_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8048072 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8048074 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8048073 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8048071 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 220479_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11746624_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11746624_at | - | - |
Human Genome U133 Plus 2.0 Array | 220479_at | - | - |
Human Genome U133 Plus 2.0 Array | 220479_PM_at | - | - |
Human Genome U133A 2.0 Array | 220479_at | - | - |
Human Genome U133A 2.0 Array | 220479_PM_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NR_002763_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw591019 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220479_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220479_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220479_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220479_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220479_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220479_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220479_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220479_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220479_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220479_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220479_at6 | - | - |
Human Genome U95 Set | 60136_PM_i_at | - | - |
Human Genome U95 Set | 60136_i_at | - | - |
Human Genome U133 Set | 220479_PM_at | - | - |
Human Genome U133 Set | 220479_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A016917 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A016917 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 132129 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1719963 | AF090897.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1719963 | NR_002763.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1719963 | AF090897.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1719963 | NR_002763.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1719963 | AF090897.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1719963 | NR_002763.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1719963 | AF090897.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1719963 | NR_002763.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1719963 | AF090897.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1719963 | NR_002763.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1719963 | AF090897.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1719963 | NR_002763.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1719963 | AF090897.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1719963 | NR_002763.2 | human_transcriptome |
