Assay Details
Gene Symbol: OPN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030015.2 | NP_001025186.1 | 5 - 6 | 887 | 90 | |
NM_033282.3 | NP_150598.1 | 4 - 5 | 854 | 90 | ||
XM_017016955.1 | XP_016872444.1 | 5 - 6 | 1949 | 90 | ||
XM_017016956.1 | XP_016872445.1 | 4 - 5 | 1916 | 90 | ||
XM_017016957.1 | XP_016872446.1 | 5 - 6 | 1684 | 90 | ||
GenBank mRNA | BC029291.1 | - | 5 - 6 | 827 | 90 | |
BC113558.1 | - | 4 - 5 | 683 | 8322610 | 90 | |
BC143688.1 | - | 5 - 6 | 716 | 9052202 | 90 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10002926.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006584.hg.1 | TC10002926.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10006584.hg.1 | ENST00000241891.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006584.hg.1 | ENST00000372071.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006584.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006584.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006585.hg.1 | TC10002926.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10006585.hg.1 | ENST00000241891.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006585.hg.1 | ENST00000372071.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006585.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006585.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156292.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156292.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156292.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156292.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156292.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156292.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156292.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156292.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156292.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156292.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156292.hg.1 | TC1000008309.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000156293.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156293.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156293.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156293.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156293.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156293.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156293.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156293.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156293.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156293.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156293.hg.1 | TC1000008309.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
