Assay Details
Gene Symbol: RGS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002927.4 | NP_002918.1 | 4 - 5 | 355 | 127 | |
GenBank mRNA | AF030107.1 | - | 4 - 5 | 363 | 127 | |
BC056866.1 | - | 4 - 5 | 354 | 4419604 | 127 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF030107.1, BC056866.1, NM_002927.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
