Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007955.hg.1 |
ENST00000329524.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007955.hg.1 |
ENST00000346183.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007955.hg.1 |
ENST00000349223.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007955.hg.1 |
ENST00000535127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007955.hg.1 |
NM_004555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007955.hg.1 |
NM_173163.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007955.hg.1 |
NM_173165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007955.hg.1 |
TC16002056.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000329524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000346183.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000349223.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000379165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000535127.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000539828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000549350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000553077.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000562926.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000563288.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000563319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000563796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000566301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000567152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000569766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000570212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000575270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
NM_004555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
NM_173163.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
NM_173165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
TC1600011417.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000329524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000346183.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000349223.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000379165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000535127.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000539828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000549350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000553077.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000562926.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000563288.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000563319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000563796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000566301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000567152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000569766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000570212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
ENST00000575270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
NM_004555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
NM_173163.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191064.hg.1 |
NM_173165.2 |
Refseq |