Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025061.hg.1 |
ENST00000391995.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025061.hg.1 |
ENST00000543215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025061.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025061.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025061.hg.1 |
TC01001623.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025063.hg.1 |
ENST00000391995.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025063.hg.1 |
ENST00000482095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025063.hg.1 |
ENST00000543215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025063.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025063.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025063.hg.1 |
TC01001623.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000482095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000482095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000482095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000482095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000482095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000482095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180809.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180809.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180809.hg.1 |
ENST00000482095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180809.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180809.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180809.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180809.hg.1 |
TC0100011014.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100180809.hg.1 |
ENST00000482095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180809.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180809.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180809.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180809.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180809.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000462955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000482095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000482095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000482095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000482095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000482095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000482095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000543215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_002927.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
NM_144766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011014.hg.1 |
ENST00000391995.6 |
Ensembl |