Assay Details
Gene Symbol: TH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199292.2 | NP_954986.2 | 2 - 3 | 203 | 57 | |
NM_199293.2 | NP_954987.2 | 2 - 3 | 191 | 57 | ||
GenBank mRNA | M17589.1 | - | 2 - 3 | 193 | 57 | |
Y00414.1 | - | 2 - 3 | 185 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
