Assay Details
Gene Symbol: COG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031431.3 | NP_113619.2 | 3 - 4 | 524 | 75 | |
XM_011535267.2 | XP_011533569.1 | 3 - 4 | 504 | 75 | ||
GenBank mRNA | AF332595.1 | - | 3 - 4 | 483 | 75 | |
AF349676.1 | - | 3 - 4 | 503 | 75 | ||
AK026305.1 | - | 3 - 4 | 483 | 75 | ||
AK225466.1 | - | 3 - 4 | 483 | 75 | ||
AK225952.1 | - | 3 - 4 | 483 | 75 | ||
AK314387.1 | - | 3 - 4 | 450 | 75 | ||
BC038953.1 | - | 3 - 4 | 414 | 5756412 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000176.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
retrograde transport, vesicle recycling within Golgi
protein glycosylation
intracellular protein transport
ER to Golgi vesicle-mediated transport
retrograde vesicle-mediated transport, Golgi to ER
intra-Golgi vesicle-mediated transport
Golgi organization
protein localization to organelle
protein stabilization
protein binding
protein transporter activity
retrograde transport, vesicle recycling within Golgi
protein glycosylation
intracellular protein transport
ER to Golgi vesicle-mediated transport
retrograde vesicle-mediated transport, Golgi to ER
intra-Golgi vesicle-mediated transport
Golgi organization
protein localization to organelle
protein stabilization
protein binding
protein transporter activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR13002033.hg.1 | ENST00000349995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13002033.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13002033.hg.1 | TC13000176.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13002032.hg.1 | ENST00000349995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13002032.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13002032.hg.1 | TC13000176.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000465942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000465942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000465942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000465942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000465942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000465942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000465942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000465942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000476702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000476702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000486940.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000486940.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000486940.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000486940.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000486940.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000486940.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000618913.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000618913.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | ENST00000618913.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300148024.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148024.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148024.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300148024.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148024.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148024.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300148024.hg.1 | TC1300007070.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000349995.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000465942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000465942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000465942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000465942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000465942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000465942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000465942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000465942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000476702.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000476702.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000486940.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000486940.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000486940.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000486940.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000486940.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000486940.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000617493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000618913.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000618913.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | ENST00000618913.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007070.hg.1 | NM_031431.3 | Refseq |
