Assay Details
Gene Symbol: C5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001317163.1 | NP_001304092.1 | 23 - 24 | 3300 | 82 | |
NM_001735.2 | NP_001726.2 | 23 - 24 | 2974 | 82 | ||
XM_011518980.2 | XP_011517282.1 | 24 - 25 | 3343 | 82 | ||
XM_017015102.1 | XP_016870591.1 | 25 - 26 | 3495 | 82 | ||
XM_017015103.1 | XP_016870592.1 | 23 - 24 | 3128 | 82 | ||
XM_017015104.1 | XP_016870593.1 | 25 - 26 | 3495 | 82 | ||
GenBank mRNA | AB209031.1 | - | 23 - 24 | 3300 | 82 | |
BC113738.1 | - | 23 - 24 | 2969 | 8322790 | 82 | |
BC113740.1 | - | 23 - 24 | 2969 | 8322792 | 82 | |
M57729.1 | - | 23 - 24 | 2956 | 82 | ||
M65134.1 | - | 13 - 14 | 1711 | 82 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001542.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
