Assay Details
Gene Symbol: ELAVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001420.3 | NP_001411.2 | 6 - 7 | 1136 | 76 | |
XM_011527778.2 | XP_011526080.1 | 6 - 7 | 1139 | 76 | ||
GenBank mRNA | AY034002.1 | - | 5 - 6 | 725 | 76 | |
BC014144.2 | - | 6 - 7 | 1129 | 76 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001185.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19016533.hg.1 | TC19001185.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19016533.hg.1 | ENST00000359227.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19016533.hg.1 | ENST00000438662.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19016533.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19016533.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000588853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000588853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000588853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000592218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000592218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | ENST00000592218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900204286.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900204286.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900204286.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900204286.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900204286.hg.1 | TC1900011870.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900204286.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900204286.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900204286.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900204286.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000588853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000588853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000588853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000592218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000592218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | ENST00000592218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011870.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900204285.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900204285.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900204285.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900204285.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900204285.hg.1 | TC1900011870.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900204285.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900204285.hg.1 | ENST00000438662.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900204285.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900204285.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900204284.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900204284.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900204284.hg.1 | NM_032281.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900204284.hg.1 | TC1900011870.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900204284.hg.1 | ENST00000359227.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900204284.hg.1 | NM_001420.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900204284.hg.1 | NM_032281.2 | Refseq |
