Assay Details
Gene Symbol: CACNA1S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000069.2 | NP_000060.2 | 39 - 40 | 5024 | 64 | |
XM_005245478.3 | XP_005245535.1 | 38 - 39 | 4854 | 64 | ||
GenBank mRNA | BC133671.1 | - | 39 - 40 | 4868 | 40073990 | 64 |
L33798.1 | - | 39 - 40 | 64 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003684.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
