Assay Details
Gene Symbol: CNKSR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173515.2 | NP_775786.2 | 9 - 10 | 1443 | 109 | |
XM_011535485.2 | XP_011533787.1 | 9 - 10 | 3926 | 109 | ||
GenBank mRNA | AK055911.1 | - | 9 - 10 | 1483 | 109 | |
AK309680.1 | - | 9 - 10 | 1049 | 109 | ||
AY328894.1 | - | 6 - 7 | 726 | 109 | ||
BC060761.1 | - | 9 - 10 | 1365 | 5299236 | 109 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06004140.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06028271.hg.1 | ENST00000367209.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06028271.hg.1 | ENST00000367213.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06028271.hg.1 | ENST00000433165.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06028271.hg.1 | ENST00000479339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06028271.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06028271.hg.1 | TC06004140.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000424998.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000424998.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000424998.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000424998.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202951.hg.1 | ENST00000424998.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202951.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202951.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202951.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202951.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202951.hg.1 | TC0600014361.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600202951.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202951.hg.1 | ENST00000424998.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202951.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202951.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202951.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000424998.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000424998.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000424998.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000424998.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
