Assay Details
Gene Symbol: BMPR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203.2 | NP_001194.1 | 2 - 3 | 169 | 77 | |
XM_017008558.1 | XP_016864047.1 | 2 - 3 | 240 | 77 | ||
GenBank mRNA | BC069796.1 | - | 1 - 2 | 81 | 7262484 | 77 |
U89326.1 | - | 2 - 3 | 168 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000503.hg
Additional Information:
This assay is in the 5'UTR region of the BC069796.1,NM_001203.2,U89326.1,XM_017008558.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
cartilage condensation
ovarian cumulus expansion
eye development
chondrocyte development
protein phosphorylation
inflammatory response
dorsal/ventral pattern formation
signal transduction by protein phosphorylation
proteoglycan biosynthetic process
positive regulation of bone mineralization
BMP signaling pathway
retinal ganglion cell axon guidance
positive regulation of chondrocyte differentiation
limb morphogenesis
ovulation cycle
positive regulation of cell differentiation
positive regulation of osteoblast differentiation
positive regulation of transcription from RNA polymerase II promoter
retina development in camera-type eye
endochondral bone morphogenesis
positive regulation of cartilage development
cellular response to BMP stimulus
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of chondrocyte proliferation
glycoprotein binding
protein serine/threonine kinase activity
transmembrane receptor protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
transforming growth factor beta receptor activity, type I
protein binding
ATP binding
SMAD binding
metal ion binding
skeletal system development
cartilage condensation
ovarian cumulus expansion
eye development
chondrocyte development
protein phosphorylation
inflammatory response
dorsal/ventral pattern formation
signal transduction by protein phosphorylation
proteoglycan biosynthetic process
positive regulation of bone mineralization
BMP signaling pathway
retinal ganglion cell axon guidance
positive regulation of chondrocyte differentiation
limb morphogenesis
ovulation cycle
positive regulation of cell differentiation
positive regulation of osteoblast differentiation
positive regulation of transcription from RNA polymerase II promoter
retina development in camera-type eye
endochondral bone morphogenesis
positive regulation of cartilage development
cellular response to BMP stimulus
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of chondrocyte proliferation
glycoprotein binding
protein serine/threonine kinase activity
transmembrane receptor protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
transforming growth factor beta receptor activity, type I
protein binding
ATP binding
SMAD binding
metal ion binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000264568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000264568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000264568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000264568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000264568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000264568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000264568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000264568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000264568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000264568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000264568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000264568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000264568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000394931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000394931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000394931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000394931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000394931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000394931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000394931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000394931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000394931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000394931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000394931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000394931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000394931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000394931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000394931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000440890.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000440890.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000440890.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000440890.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000440890.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000440890.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000440890.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000440890.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000440890.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000440890.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000440890.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000440890.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000440890.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000440890.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000440890.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000440890.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000502683.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000502683.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000502683.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000502683.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000502683.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000506363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000506363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000506363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000506363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000506363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000506363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000509540.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000509540.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000509540.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000509540.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000509540.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000509540.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000509540.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000509540.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000509540.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000509540.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000509540.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000509540.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000509540.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000512312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000512312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000512312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000512312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000512312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000512312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000512312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000512312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000512312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000512312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000512312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000512312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | ENST00000515059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001203.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256792.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256792.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256792.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256792.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256792.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256792.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256792.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256792.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256792.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256792.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256792.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256792.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256792.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256792.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256792.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256792.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256794.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256794.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256794.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256794.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256794.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256794.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256794.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256794.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256794.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256794.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256794.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256794.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256794.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256794.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256794.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008183.hg.1 | NM_001256794.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 188925 | - | - |
