Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000264568.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000264568.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000264568.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000264568.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000264568.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000264568.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000264568.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000264568.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000264568.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000264568.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000264568.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000264568.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000264568.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000394931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000394931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000394931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000394931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000394931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000394931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000394931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000394931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000394931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000394931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000394931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000394931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000394931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000394931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000394931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000440890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000440890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000440890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000440890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000440890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000440890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000440890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000440890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000440890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000440890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000440890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000440890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000440890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000440890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000440890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000440890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000502683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000502683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000502683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000502683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000502683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000506363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000506363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000506363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000506363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000506363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000506363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000509540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000509540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000509540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000509540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000509540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000509540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000509540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000509540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000509540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000509540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000509540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000509540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000509540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000512312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000512312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000512312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000512312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000512312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000512312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000512312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000512312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000512312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000512312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000512312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000512312.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
ENST00000515059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001203.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256792.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008183.hg.1 |
NM_001256794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156333.hg.1 |
ENST00000394931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156333.hg.1 |
ENST00000394931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156333.hg.1 |
TC0400008183.hg.1 |
Clariom D TC |