Assay Details
Gene Symbol: NPFFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004885.2 | NP_004876.2 | 1 - 2 | 398 | 94 | |
GenBank mRNA | AF119815.1 | - | 1 - 2 | 398 | 94 | |
AK308069.1 | - | 1 - 2 | 389 | 94 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000397.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
G protein-coupled receptor signaling pathway
neuropeptide signaling pathway
chemical synaptic transmission
detection of abiotic stimulus
cellular response to hormone stimulus
regulation of MAPK cascade
regulation of adenylate cyclase activity
response to peptide
regulation of cAMP-dependent protein kinase activity
G protein-coupled receptor activity
neuropeptide receptor activity
opioid receptor binding
peptide binding
G protein-coupled receptor signaling pathway
neuropeptide signaling pathway
chemical synaptic transmission
detection of abiotic stimulus
cellular response to hormone stimulus
regulation of MAPK cascade
regulation of adenylate cyclase activity
response to peptide
regulation of cAMP-dependent protein kinase activity
G protein-coupled receptor activity
neuropeptide receptor activity
opioid receptor binding
peptide binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04005476.hg.1 | ENST00000308744.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005476.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04005476.hg.1 | TC04000397.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000344413.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000344413.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000344413.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000358749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000358749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000358749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000358749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000358749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000358749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000395999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000395999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000395999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000395999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000395999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000395999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000395999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000395999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000506359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000506359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000506359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | ENST00000506359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04005477.hg.1 | ENST00000308744.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005477.hg.1 | ENST00000344413.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005477.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04005477.hg.1 | TC04000397.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000344413.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000344413.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000344413.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000358749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000358749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000358749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000358749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000358749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000358749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000395999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000395999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000395999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000395999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000395999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000395999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000395999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000395999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000506359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000506359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000506359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | ENST00000506359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_001144756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007806.hg.1 | NM_053036.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153425.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153425.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153425.hg.1 | TC0400007806.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400153425.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153425.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153426.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153426.hg.1 | ENST00000344413.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153426.hg.1 | NM_004885.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153426.hg.1 | TC0400007806.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400153426.hg.1 | ENST00000308744.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153426.hg.1 | ENST00000344413.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153426.hg.1 | NM_004885.2 | Refseq |
