Assay Details
Gene Symbol: GRIK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000831.3 | NP_000822.2 | 9 - 10 | 1465 | 85 | |
GenBank mRNA | AJ249210.1 | - | 9 - 10 | 1351 | 85 | |
AJ299451.1 | - | 9 - 10 | 1330 | 85 | ||
U16127.1 | - | 9 - 10 | 1348 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002513.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
adenylate cyclase-inhibiting G protein-coupled glutamate receptor signaling pathway
glutamate receptor signaling pathway
G protein-coupled glutamate receptor signaling pathway
ion transmembrane transport
ionotropic glutamate receptor signaling pathway
regulation of membrane potential
negative regulation of synaptic transmission, glutamatergic
adenylate cyclase inhibiting G protein-coupled glutamate receptor activity
G protein-coupled receptor binding
ionotropic glutamate receptor activity
extracellularly glutamate-gated ion channel activity
glutamate receptor activity
ligand-gated ion channel activity
kainate selective glutamate receptor activity
adenylate cyclase-inhibiting G protein-coupled glutamate receptor signaling pathway
glutamate receptor signaling pathway
G protein-coupled glutamate receptor signaling pathway
ion transmembrane transport
ionotropic glutamate receptor signaling pathway
regulation of membrane potential
negative regulation of synaptic transmission, glutamatergic
adenylate cyclase inhibiting G protein-coupled glutamate receptor activity
G protein-coupled receptor binding
ionotropic glutamate receptor activity
extracellularly glutamate-gated ion channel activity
glutamate receptor activity
ligand-gated ion channel activity
kainate selective glutamate receptor activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01039045.hg.1 | ENST00000373091.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039045.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039045.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039045.hg.1 | TC01002513.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000479620.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000479620.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000479620.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000479620.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000479620.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000479620.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000479620.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000479620.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100202403.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100202403.hg.1 | ENST00000373093.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100202403.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100202403.hg.1 | TC0100013730.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100202403.hg.1 | NM_000831.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100202403.hg.1 | ENST00000373091.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100202403.hg.1 | ENST00000373093.4 | Ensembl |
