Assay Details
Gene Symbol: GABRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000812.3 | NP_000803.2 | 1 - 2 | 457 | 73 | |
XM_017007986.1 | XP_016863475.1 | 1 - 2 | 182 | 73 | ||
GenBank mRNA | AK225167.1 | - | 1 - 2 | 134 | 73 | |
AK225686.1 | - | 1 - 2 | 161 | 73 | ||
AK293691.1 | - | 1 - 2 | 73 | |||
AK296023.1 | - | 1 - 2 | 350 | 73 | ||
AK307120.1 | - | 1 - 2 | 184 | 73 | ||
AK312720.1 | - | 1 - 2 | 103 | 73 | ||
BC022449.1 | - | 1 - 2 | 161 | 4797401 | 73 | |
DC352362.1 | - | 1 - 2 | 457 | 73 | ||
KJ534841.1 | - | 1 - 2 | 83 | 73 | ||
KJ535054.1 | - | 1 - 2 | 83 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000273.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK293691.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transport
ion transport
signal transduction
response to toxic substance
central nervous system neuron development
response to progesterone
ion transmembrane transport
ovulation cycle
cellular response to histamine
chloride transmembrane transport
GABA-A receptor activity
ligand-gated ion channel activity
GABA-gated chloride ion channel activity
GABA receptor binding
transport
ion transport
signal transduction
response to toxic substance
central nervous system neuron development
response to progesterone
ion transmembrane transport
ovulation cycle
cellular response to histamine
chloride transmembrane transport
GABA-A receptor activity
ligand-gated ion channel activity
GABA-gated chloride ion channel activity
GABA receptor binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04003916.hg.1 | ENST00000295454.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04003916.hg.1 | ENST00000509366.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04003916.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04003916.hg.1 | TC04000273.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04003912.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04003912.hg.1 | TC04000273.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04003913.hg.1 | ENST00000295454.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04003913.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04003913.hg.1 | TC04000273.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04003917.hg.1 | ENST00000295454.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04003917.hg.1 | ENST00000509366.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04003917.hg.1 | ENST00000538619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04003917.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04003917.hg.1 | TC04000273.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04003918.hg.1 | ENST00000295454.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04003918.hg.1 | ENST00000509366.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04003918.hg.1 | ENST00000538619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04003918.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04003918.hg.1 | TC04000273.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000381582.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000381582.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000381582.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000381582.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000381582.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000509366.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000509366.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000509366.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000509366.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000509366.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000510909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000510909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000510909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000510909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000510909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000513567.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000513567.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000513567.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | ENST00000513567.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400150658.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150658.hg.1 | ENST00000381582.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150658.hg.1 | ENST00000509366.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150658.hg.1 | ENST00000510909.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150658.hg.1 | ENST00000513567.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150658.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400150658.hg.1 | TC0400007419.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400150658.hg.1 | ENST00000509366.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150658.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150658.hg.1 | ENST00000381582.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150658.hg.1 | ENST00000510909.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150658.hg.1 | ENST00000513567.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150658.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400150651.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400150651.hg.1 | TC0400007419.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400150651.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400150652.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150652.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400150652.hg.1 | TC0400007419.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400150652.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150652.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400150655.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150655.hg.1 | ENST00000381582.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150655.hg.1 | ENST00000509366.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150655.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400150655.hg.1 | TC0400007419.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400150655.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150655.hg.1 | ENST00000381582.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150655.hg.1 | ENST00000509366.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150655.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000381582.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000381582.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000381582.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000381582.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000381582.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000509366.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000509366.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000509366.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000509366.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000509366.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000510909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000510909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000510909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000510909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000510909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000513567.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000513567.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000513567.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | ENST00000513567.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007419.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400150657.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150657.hg.1 | ENST00000381582.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150657.hg.1 | ENST00000509366.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150657.hg.1 | ENST00000510909.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150657.hg.1 | NM_000812.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400150657.hg.1 | TC0400007419.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400150657.hg.1 | ENST00000295454.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150657.hg.1 | ENST00000381582.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150657.hg.1 | ENST00000509366.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150657.hg.1 | ENST00000510909.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150657.hg.1 | NM_000812.3 | Refseq |
