Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015971.hg.1 |
ENST00000302930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015971.hg.1 |
ENST00000355591.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015971.hg.1 |
ENST00000396533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015971.hg.1 |
ENST00000543208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015971.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015971.hg.1 |
TC04001164.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015976.hg.1 |
ENST00000355591.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015976.hg.1 |
ENST00000396533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015976.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015976.hg.1 |
TC04001164.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015970.hg.1 |
ENST00000302930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015970.hg.1 |
ENST00000355591.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015970.hg.1 |
ENST00000396533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015970.hg.1 |
ENST00000543208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015970.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015970.hg.1 |
TC04001164.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015969.hg.1 |
ENST00000302930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015969.hg.1 |
ENST00000355591.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015969.hg.1 |
ENST00000396533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015969.hg.1 |
ENST00000543208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015969.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015969.hg.1 |
TC04001164.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010601.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172234.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172234.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172234.hg.1 |
TC0400010601.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400172234.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172234.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172233.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172233.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172233.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172233.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172233.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172233.hg.1 |
TC0400010601.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400172233.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172233.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172233.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172233.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172233.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172237.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172237.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172237.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172237.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172237.hg.1 |
TC0400010601.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400172237.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172237.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172237.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172237.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172231.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172231.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172231.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172231.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172231.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172231.hg.1 |
TC0400010601.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400172231.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172231.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172231.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172231.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172231.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172232.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172232.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172232.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172232.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172232.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172232.hg.1 |
TC0400010601.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400172232.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172232.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172232.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172232.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172232.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000355591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000396533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000505102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
ENST00000543208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
NM_130902.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010601.hg.1 |
NM_130902.2 |
Refseq |