Assay Details
Gene Symbol: CYP27A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000784.3 | NP_000775.1 | 7 - 8 | 1701 | 96 | |
XM_017003488.1 | XP_016858977.1 | 6 - 7 | 2253 | 96 | ||
GenBank mRNA | AK290418.1 | - | 7 - 8 | 1307 | 96 | |
BC040430.1 | - | 7 - 8 | 1281 | 5760376 | 96 | |
BC051851.1 | - | 7 - 8 | 1279 | 5240515 | 96 | |
M62401.1 | - | 7 - 8 | 1288 | 96 | ||
X59812.1 | - | 7 - 8 | 1464 | 96 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001311.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02020306.hg.1 | ENST00000258415.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02020306.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02020306.hg.1 | TC02001311.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02020312.hg.1 | ENST00000258415.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02020312.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02020312.hg.1 | TC02001311.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02020308.hg.1 | ENST00000258415.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02020308.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02020308.hg.1 | TC02001311.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02020310.hg.1 | ENST00000258415.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02020310.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02020310.hg.1 | TC02001311.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0200177156.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177156.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177156.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200177156.hg.1 | TC0200010811.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200177156.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200177156.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177156.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177158.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177158.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177158.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200177158.hg.1 | TC0200010811.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200177158.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177158.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177158.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200177152.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177152.hg.1 | ENST00000445971.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177152.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177152.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177152.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200177152.hg.1 | TC0200010811.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200177152.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177152.hg.1 | ENST00000445971.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177152.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177152.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177152.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200177154.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177154.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177154.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200177154.hg.1 | TC0200010811.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200177154.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177154.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200177154.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
