Assay Details
Gene Symbol: S1PR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005226.3 | NP_005217.2 | 1 - 2 | 293 | 74 | |
GenBank mRNA | AK312798.1 | - | 1 - 2 | 257 | 74 | |
BC060827.1 | - | 1 - 2 | 255 | 30342032 | 74 | |
DA722994.1 | - | 1 - 2 | 293 | 74 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK312798.1, BC060827.1, NM_005226.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
sphingosine-1-phosphate receptor signaling pathway
inflammatory response
G protein-coupled receptor signaling pathway
adenylate cyclase-inhibiting G protein-coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
Notch signaling pathway
positive regulation of cell population proliferation
anatomical structure morphogenesis
regulation of interleukin-1 beta production
negative regulation of establishment of endothelial barrier
G protein-coupled receptor activity
integrin binding
protein binding
lipid binding
sphingosine-1-phosphate receptor activity
cytokine production
sphingosine-1-phosphate receptor signaling pathway
inflammatory response
G protein-coupled receptor signaling pathway
adenylate cyclase-inhibiting G protein-coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
Notch signaling pathway
positive regulation of cell population proliferation
anatomical structure morphogenesis
regulation of interleukin-1 beta production
negative regulation of establishment of endothelial barrier
G protein-coupled receptor activity
integrin binding
protein binding
lipid binding
sphingosine-1-phosphate receptor activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000334490.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000334490.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000334490.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000334490.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000334490.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000334490.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000334490.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000334490.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000358157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000358157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000358157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000358157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375846.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375846.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375846.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375846.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375850.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375850.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375850.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375850.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375850.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375850.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375851.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375851.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375851.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375851.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375851.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375851.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375851.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | ENST00000375851.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_001142413.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_001142413.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_001142413.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_001142413.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_001142413.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_001142413.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_001142413.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_001142413.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_005226.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_005226.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_005226.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_005226.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_005226.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007854.hg.1 | NM_005226.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900152991.hg.1 | ENST00000358157.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900152991.hg.1 | ENST00000375846.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900152991.hg.1 | NM_005226.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900152991.hg.1 | TC0900007854.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900152991.hg.1 | NM_005226.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900152991.hg.1 | ENST00000358157.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900152991.hg.1 | ENST00000375846.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900152992.hg.1 | ENST00000358157.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900152992.hg.1 | ENST00000375846.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900152992.hg.1 | NM_005226.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900152992.hg.1 | TC0900007854.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900152992.hg.1 | ENST00000358157.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900152992.hg.1 | ENST00000375846.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900152992.hg.1 | NM_005226.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900152981.hg.1 | NM_005226.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900152981.hg.1 | TC0900007854.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900152981.hg.1 | NM_005226.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000334490.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000334490.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000334490.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000334490.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000334490.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000334490.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000334490.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000334490.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000358157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000358157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000358157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000358157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375846.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375846.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375846.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375846.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375850.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375850.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375850.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375850.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375850.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375850.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375851.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375851.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375851.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375851.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375851.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375851.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375851.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | ENST00000375851.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_001001938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_001142413.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_001142413.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_001142413.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_001142413.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_001142413.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_001142413.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_001142413.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_001142413.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_005226.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_005226.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_005226.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_005226.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_005226.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007854.hg.1 | NM_005226.3 | Refseq |
