Assay Details
Gene Symbol: LOC283911 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL833480.1 | - | 1 | 609 | 57 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | putative cyclin-Y-like protein 3 | LOC102724928 | -88.3kb | Sense | Chr. 16 - 35030347 - 35060329 on NCBI Build GRCh38 | |
Downstream | ubiquitin conjugating enzyme E2 M pseudogene 1 | UBE2MP1 | 20.1kb | Anti-sense | Chr. 16 - 35169431 - 35170391 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988786 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988788 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988877 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527381 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988802 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988819 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988801 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988823 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988832 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988813 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988805 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988881 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988790 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527372 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16890706 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527305 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988874 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988848 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988783 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988821 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988869 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988876 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527307 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988811 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988837 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988882 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527342 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988843 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988827 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988826 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988807 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988782 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16890699 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988780 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988860 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988787 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988814 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988795 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988824 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988878 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988822 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988846 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527362 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527326 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988835 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527312 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527348 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988789 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988818 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988870 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988841 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988868 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527340 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527373 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988854 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527378 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16890712 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988791 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988817 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527306 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988836 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988800 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988804 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527310 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988851 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988820 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988798 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527364 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988830 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988840 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988863 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527360 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527389 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988793 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988872 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527391 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988857 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527398 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988880 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988866 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988785 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988803 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988859 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988839 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988815 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988865 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30527328 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 16523 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 23265 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 226910 | AB023218.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 226910 | AY358380.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 226910 | BC012375.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 226910 | CR593524.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 226910 | NM_014960.2 | refseq_mrna_human_NM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 523118 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 265988 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3808371 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P123844 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P123844 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 47250 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P20451220 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3808371 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P123844 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P123844 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P123844 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 161023 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 16782 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P123844 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03136664 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P416525 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03136664 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P123844 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600010132.hg.1 | ENST00000563142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007662.hg.1 | ENST00000623041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007662.hg.1 | ENST00000623041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010132.hg.1 | ENST00000563142.1 | Ensembl |
