Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000368923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000368923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000368923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000368923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000368923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000368923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000479373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000479373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000479373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000479373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000479373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
ENST00000479373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
NM_004032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
NM_004032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
NM_004032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
NM_004032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012832.hg.1 |
NM_004032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025039.hg.1 |
TC06004136.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025039.hg.1 |
ENST00000368923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025039.hg.1 |
ENST00000368924.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025039.hg.1 |
ENST00000368925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025039.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025039.hg.1 |
NM_004032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025041.hg.1 |
TC06004136.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025041.hg.1 |
ENST00000368923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025041.hg.1 |
ENST00000368924.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025041.hg.1 |
ENST00000368925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025041.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025041.hg.1 |
NM_004032.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188602.hg.1 |
ENST00000368923.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188602.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188602.hg.1 |
ENST00000479373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188602.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188602.hg.1 |
NM_004032.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188602.hg.1 |
ENST00000368923.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188602.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188602.hg.1 |
ENST00000479373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188602.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188602.hg.1 |
NM_004032.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188602.hg.1 |
TC0600012832.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000368923.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000368923.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000368923.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000368923.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000368923.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000368923.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000479373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000479373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000479373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000479373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000479373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
ENST00000479373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
NM_004032.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
NM_004032.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
NM_004032.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
NM_004032.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012832.hg.1 |
NM_004032.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188603.hg.1 |
ENST00000368923.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188603.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188603.hg.1 |
ENST00000479373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188603.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188603.hg.1 |
NM_004032.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188603.hg.1 |
ENST00000368923.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188603.hg.1 |
ENST00000368924.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188603.hg.1 |
ENST00000479373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188603.hg.1 |
NM_003649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188603.hg.1 |
NM_004032.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188603.hg.1 |
TC0600012832.hg.1 |
Clariom D TC |