Assay Details
Gene Symbol: MTMR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306144.1 | NP_001293073.1 | 2 - 3 | 418 | 140 | |
GenBank mRNA | AK226109.1 | - | 2 - 3 | 372 | 140 | |
AK295738.1 | - | 2 - 3 | 213 | 140 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK295738.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
