Assay Details
Gene Symbol: MOXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015529.3 | NP_056344.2 | 1 - 2 | 366 | 141 | |
GenBank mRNA | AK074879.1 | - | 1 - 2 | 81 | 141 | |
AY359094.1 | - | 1 - 2 | 290 | 141 | ||
BC018756.1 | - | 1 - 2 | 325 | 4841330 | 141 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06002113.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06026523.hg.1 | TC06002113.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06026523.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06026523.hg.1 | ENST00000367963.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06026523.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06026522.hg.1 | TC06002113.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06026522.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06026522.hg.1 | ENST00000367963.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06026522.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000392401.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000392401.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000392401.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06026526.hg.1 | TC06002113.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06026526.hg.1 | ENST00000367963.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06026526.hg.1 | ENST00000392401.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06026526.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06026528.hg.1 | TC06002113.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06026528.hg.1 | ENST00000367963.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06026528.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191068.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191068.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191068.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191068.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191068.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191068.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191068.hg.1 | TC0600013190.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600191072.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191072.hg.1 | ENST00000392401.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191072.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191072.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191072.hg.1 | ENST00000392401.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191072.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191072.hg.1 | TC0600013190.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000392401.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000392401.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000392401.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191073.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191073.hg.1 | ENST00000392401.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191073.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191073.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191073.hg.1 | ENST00000392401.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191073.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191073.hg.1 | TC0600013190.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600191069.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191069.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191069.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191069.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191069.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191069.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191069.hg.1 | TC0600013190.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600191075.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191075.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191075.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191075.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191075.hg.1 | TC0600013190.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 105667 | - | - |
